Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLASP1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106584 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-584 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-584 Supplier Abnova Corporation Supplier No. H00023332A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CLASP1.

CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs (e.g., CLIP170; MIM 179838). CLASP1 is involved in the regulation of microtubule dynamics at the kinetochore and throughout the spindle (Maiato et al., 2003 [PubMed 12837247]).[supplied by OMIM

Sequence: DGLAKHPPPFSQPNSIPTAPSHKALRRSYSPSMLDYDTENLNSEEIYSSLRGVTEAIEKFSFRSQEDLNEPIKRDGKKECDIVSRDGGAASPAT

Specifications

Antigen CLASP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CLASP1.
Formulation 50% glycerol
Gene CLASP1
Gene Accession No. NM_015282
Gene Alias DKFZp686D1968/DKFZp686H2039/FLJ33821/FLJ41222/KIAA0622/MAST1/MGC131895
Gene Symbols CLASP1
Host Species Mouse
Immunogen CLASP1 (NP_056097, 1133 a.a. to 1226 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23332
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.