Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLCA2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106128 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-128 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-128 Supplier Abnova Corporation Supplier No. H00009635A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CLCA2.

The protein encoded by this gene belongs to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. Since this protein is expressed predominantly in trachea and lung, it is suggested to play a role in the complex pathogenesis of cystic fibrosis. It may also serve as adhesion molecule for lung metastatic cancer cells, mediating vascular arrest and colonization, and furthermore, it has been implicated to act as a tumor suppressor gene for breast cancer. [provided by RefSeq

Sequence: PTFSLVQAGDKVVCLVLDVSSKMAEADRLLQLQQAAEFYLMQIVEIHTFVGIASFDSKGEIRAQLHQINSNDDRKLLVSYLPTTVSAKTDISICSGLKKGF

Specifications

Antigen CLCA2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CLCA2.
Formulation 50% glycerol
Gene CLCA2
Gene Accession No. NM_006536
Gene Alias CACC/CACC3/CLCRG2/CaCC-3/FLJ97885
Gene Symbols CLCA2
Host Species Mouse
Immunogen CLCA2 (NP_006527, 300 a.a. to 400 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9635
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.