Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLDN19, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107649 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-649 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-649 Supplier Abnova Corporation Supplier No. H00149461A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant CLDN19.

The product of this gene belongs to the claudin family. It plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity. Defects in this gene are the cause of hypomagnesemia renal with ocular involvement (HOMGO). HOMGO is a progressive renal disease characterized by primary renal magnesium wasting with hypomagnesemia, hypercalciuria and nephrocalcinosis associated with severe ocular abnormalities such as bilateral chorioretinal scars, macular colobomata, significant myopia and nystagmus. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MANSGLQLLGYFLALGGWVGIIASTALPQWKQSSYAGDAIITAVGLYEGLWMSCASQSTGQVQCKLYDSLLALDGHIQSARALMVVAVLLGFVAMVLSVVGMKCTRVGDSNPIAKGRVAIAGGALFILAGLCTLTAVSWYATLVTQEFFNPSTPVNARYEFGPALFVGWASAGLAVLGGSFLCCTCPEPERPNSSPQPYRPGPSAAAREYV

Specifications

Antigen CLDN19
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant CLDN19.
Formulation 50% glycerol
Gene CLDN19
Gene Accession No. BC030524
Gene Symbols CLDN19
Host Species Mouse
Immunogen CLDN19 (AAH30524, 1 a.a. to 211 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149461
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.