Learn More
CLEC2B, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CLEC2B.
Supplier: Abnova Corporation H00009976A01
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell activation antigen. An alternative splice variant has been described but its full-length sequence has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region. [provided by RefSeq
Sequence: EEGDWNSSKYNCSTQHADLTIIDNIEEMNFLRRYKCSSDHWIGLKMAKNRTGQWVDGATFTKSFGMRGSEGCAYLSDDGAATARCYTERKWICRKRIHSpecifications
CLEC2B | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CLEC2B. | |
CLEC2B | |
AICL/CLECSF2/HP10085/IFNRG1 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_005127 | |
CLEC2B | |
CLEC2B (NP_005118, 52 a.a. to 149 a.a) partial recombinant protein with GST tag. | |
RUO | |
9976 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only