Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLU, Mouse, Clone: 16, Abnova™

Catalog No. 89113920 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-920 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-920 Supplier Abnova Corporation Supplier No. MAB9962
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CLU.

Specifications

Antigen CLU
Applications ELISA, Western Blot
Classification Monoclonal
Clone 16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CLU.
Dilution Western blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene CLU
Gene Alias AAG4/APOJ/CLI/KUB1/MGC24903/SGP-2/SGP2/SP-40/TRPM-2/TRPM2
Gene Symbols CLU
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 23-449 of human CLU.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1191
Test Specificity It can expression in H520 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.