Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLU, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104293 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-293 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-293 Supplier Abnova Corporation Supplier No. H00001191A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CLU.

Sequence: WKMLNTSSLLEQLNEQFNWVSRLANLTQGEDQYYLRVTTVASHTSDSDVPSGVTEVVVKLFDSDPITVTVPVEVSRKNPKFMETVAEKALQEYRKKHREE

Specifications

Antigen CLU
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CLU.
Formulation 50% glycerol
Gene CLU
Gene Accession No. NM_001831
Gene Alias AAG4/APOJ/CLI/KUB1/MGC24903/SGP-2/SGP2/SP-40/TRPM-2/TRPM2
Gene Symbols CLU
Host Species Mouse
Immunogen CLU (NP_001822, 402 a.a. to 501 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1191
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.