Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMBL, Mouse, Clone: AT2B11, Abnova™

Catalog No. 89109367 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-367 Supplier Abnova Corporation Supplier No. MAB11199
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CMBL.

Specifications

Antigen CMBL
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT2B11
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CMBL.
Dilution Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) with 0.09% sodium azide.
Gene CMBL
Gene Alias FLJ23617
Gene Symbols CMBL
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-245 of human CMBL.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134147
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.