Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CML, Mouse, Biotin, Clone: NF-1G, Abnova™

Catalog No. 89136243 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-243 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-243 Supplier Abnova Corporation Supplier No. MAB6592
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant CML.

Specifications

Antigen CML
Applications Immunohistochemistry
Classification Monoclonal
Clone NF-1G
Conjugate Biotin
Description Mouse monoclonal antibody raised against recombinant CML.
Dilution Immunohistochemistry (3-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In serum (2% Block Ace, 0.1% proclin)
Host Species Mouse
Immunogen Recombinant protein (conjugated with HSA) corresponding to CML.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.