Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMTM5, Mouse, Clone: 2B10, Abnova™

Catalog No. 89107565 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-565 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-565 Supplier Abnova Corporation Supplier No. H00116173M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CMTM5.

This gene belongs to the chemokine-like factor gene superfamily. This family of genes encodes multi-pass membrane proteins that are similar to both the chemokine and the transmembrane 4 superfamilies of signaling molecules. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: MLSARDRRDRHPEEGVVAELQGFAVDKAFLTSHKGILLETELALTLIIFICFTASISAYMAAALLEFFITLAFLFLYATQYYQRFDRINWPCLDFLRCVSAIIIFLVVSFAAVTSRDGAAIAAFVFGIILVSIFAYDAFKIYRTEMAPGASQGDQQ

Specifications

Antigen CMTM5
Applications ELISA
Classification Monoclonal
Clone 2B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CMTM5.
Formulation ascites with no preservative
Gene CMTM5
Gene Accession No. BC013109
Gene Alias CKLFSF5/FLJ37521
Gene Symbols CMTM5
Host Species Mouse
Immunogen CMTM5 (AAH13109,1 a.a. ∽ 156 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116173
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.