Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMTM7, Mouse, Clone: S1, Abnova™

Catalog No. 89107529 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-529 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-529 Supplier Abnova Corporation Supplier No. H00112616M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CMTM7.

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. The protein encoded by this gene is highly expressed in leukocytes, but its exact function is unknown. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: MSHGAGLVRTTCSSGSALGPGAGAAQPSASPLEGLLDLSYPRTHAALLKVAQMVTLLIAFICVRSSLWTNYSAYSYFEVVTICDLIMILAFYLVHLFRFYRVLTCISWPLSELLHYLIGTLLLLIASIVAASKSYNQSGLVAGAIFGFMATFLCMASIWLSYKISCVTQSTDAAV

Specifications

Antigen CMTM7
Applications ELISA
Classification Monoclonal
Clone S1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CMTM7.
Formulation ascites with no preservative
Gene CMTM7
Gene Accession No. BC010116.1
Gene Alias CKLFSF7/FLJ30992
Gene Symbols CMTM7
Host Species Mouse
Immunogen CMTM7 (AAH10116.1,1 a.a. ∽ 175 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112616
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.