Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cntn1, Mouse, Biotin, Clone: S73-20, Abnova™

Catalog No. 89110095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-095 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-095 Supplier Abnova Corporation Supplier No. MAB11955

Mouse monoclonal antibody raised against synthetic peptide of Cntn1.

Sequence: EFTWHRRYGHGVSEEDKC

Specifications

Antigen Cntn1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S73-20
Conjugate Biotin
Description Mouse monoclonal antibody raised against synthetic peptide of Cntn1.
Dilution Western Blot (1 ug/mL) Immunocytochemistry (0.1-1 ug/mL) Immunofluorescence (1-10 ug/mL) Immunohistochemistry (0.1-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Cntn1
Gene Alias F3
Gene Symbols Cntn1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids of 21-37 at extracellular domain of rat Cntn1.
Purification Method Protein G Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117258
Target Species Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.