Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL16A1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104308 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-308 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-308 Supplier Abnova Corporation Supplier No. H00001307A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant COL16A1.

This gene encodes the alpha chain of type XVI collagen, a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Members of this collagen family are found in association with fibril-forming collagens such as type I and II, and serve to maintain the integrity of the extracellular matrix. High levels of type XVI collagen have been found in fibroblasts and keratinocytes, and in smooth muscle and amnion. [provided by RefSeq

Sequence: WYLFQVTDANGYPQISLEVNSQERSLELRAQGQDGDFVSCIFPVPQLFDLRWHKLMLSVAGRVASVHVDCSSASSQPLGPRRPMRPVGHVFLGLDAEQGK

Specifications

Antigen COL16A1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant COL16A1.
Formulation 50% glycerol
Gene COL16A1
Gene Accession No. NM_001856
Gene Alias 447AA/FP1572
Gene Symbols COL16A1
Host Species Mouse
Immunogen COL16A1 (NP_001847, 117 a.a. to 216 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1307
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.