Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL18A1, Mouse, Clone: KT58, Abnova™

Catalog No. 89113747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-747 Supplier Abnova Corporation Supplier No. MAB9727
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant COL18A1.

This gene encodes the alpha chain of type XVIII collagen. This collagen is one of the multiplexins, extracellular matrix proteins that contain multiple triple-helix domains (collagenous domains) interrupted by noncollagenous domains. The proteolytically produced C-terminal fragment of type XVIII collagen is endostatin, a potent antiangiogenic protein. Mutations in this gene are associated with Knobloch syndrome. The main features of this syndrome involve retinal abnormalities, so type XVIII collagen may play an important role in retinal structure and in neural tube closure. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen COL18A1
Applications ELISA
Classification Monoclonal
Clone KT58
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant COL18A1.
Dilution ELISA (1 ug/mL) Sandwich ELISA (5 ug/mL for coating) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene COL18A1
Gene Alias FLJ27325/FLJ34914/KNO/KNO1/MGC74745
Gene Symbols COL18A1
Host Species Mouse
Immunogen Recombinant protein corresponding to human COL18A1.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80781
Target Species Human
Content And Storage Store at 4°C for 1 month. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.