Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL3A1, Mouse, Clone: FH-7A, Abnova™

Catalog No. 89113145 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-145 Supplier Abnova Corporation Supplier No. MAB7205

Mouse monoclonal antibody raised against full-length native COL3A1.

This gene encodes the pro-alpha1 chains of type III collagen, a fibrillar collagen that is found in extensible connective tissues such as skin, lung, uterus, intestine and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome types IV, and with aortic and arterial aneurysms. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene. [provided by R. Dalgleish

Specifications

Antigen COL3A1
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone FH-7A
Conjugate Unconjugated
Description collagen, type III, alpha 1
Formulation In ascites (0.09% sodium azide)
Gene COL3A1
Gene Alias EDS4A, FLJ34534
Gene Symbols COL3A1
Host Species Mouse
Immunogen Native purified human COL3A1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1281
Content And Storage Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.