Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV, Mouse, Clone: 1042, Abnova™

Catalog No. 89135933 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-933 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-933 Supplier Abnova Corporation Supplier No. MAB5310

Mouse monoclonal antibody raised against native Collagen IV.

Specifications

Antigen Collagen IV
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1042
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Collagen IV.
Dilution Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation In 150mM PBS (1% BSA, 0.02% sodium azide)
Host Species Mouse
Immunogen Native purified from human placenta extracellular basement membranes.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody reacts exclusively with human collagen type IV. In immunoblots This antibody is exclusively reactive with basal membranes. There is no crossreactivity with any other collagen types.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.