Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV, Mouse, Clone: COL 94, Abnova™

Catalog No. 89135449 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-449 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-449 Supplier Abnova Corporation Supplier No. MAB3532
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Collagen IV.

Specifications

Antigen Collagen IV
Applications Immunohistochemistry
Classification Monoclonal
Clone COL 94
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Collagen IV.
Dilution Immunohistochemistry (1:10-1:50) The optimal working dilution should be determined by the end user.
Formulation In phosphate buffer, pH 7.3 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified human Collagen IV.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with human collagen IV, no cross-reactivity with other collagen types, Vitronectin, Fibronectin or Chondroitinsulfate.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.