Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COMP, Mouse, Clone: 6G5, Abnova™

Catalog No. 89111580 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-580 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-580 Supplier Abnova Corporation Supplier No. MAB3751
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against COMP.

The protein encoded by this gene is a noncollagenous extracellular matrix (ECM) protein. It consists of five identical glycoprotein subunits, each with EGF-like and calcium-binding (thrombospondin-like) domains. Oligomerization results from formation of a five-stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED). [provided by RefSeq

Specifications

Antigen COMP
Applications ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against COMP.
Dilution ELISA (1:5-1:10) Western Blot (1:1-1:3) The optimal working dilution should be determined by the end user.
Formulation In culture supernatant (0.01% sodium azide)
Gene COMP
Gene Alias EDM1/EPD1/MED/MGC131819/MGC149768/PSACH/THBS5
Gene Symbols COMP
Host Species Mouse
Immunogen Native purified COMP from normal human cartilage.
Quantity 10 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1311
Test Specificity Reacts with both reduced and non-reduced human COMP.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.