Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Complement C3b/iC3b/C3dg, Clone: 1H8,

Catalog No. MABF983MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF983MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF983MI Supplier MilliporeSigma™ Supplier No. MABF983
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Complement C3b/iC3b/C3dg clone: 1H8 in Human, Primate samples, and it is validated for Flow Cytometry

Complement C3 (UniProt P01024; also known as C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1) is encoded by the C3 (also known as CPAMD1) gene (Gene ID 718) in human. C3 is initially translated with an N-terminal 22-amino acid signal peptide sequence, which is then removed to produce the 1641-amino acid mature C3. It plays a central role in the activation of complement system. Its activation is required for the activation of both classical and alternative pathway of complement (CPC and APC, respectively). C3 is cleaved into C3a and C3b during CPC activation by the C3-convertase C4b2a composed of the activated C4 and C2. In APC, C3 is cleaved by the C3-convertase C3bBb composed C3b and the activated form of factor B (Bb). C3b serves as an opsonizing agent, and can be further cleaved by Factor I into C3c and C3d. iC3b is a proteolytically inactive C3b fragment that still opsonizes target microbes or cells, but cannot further amplify/activate the complement cascade through APC. iC3b can be further cleaved to C3dg, and finally to C3d. Unregulated activation of APC can result in paroxysmal nocturnal hemoglobinuria (PNH) that is characterized by chronic intravascular hemolysis. Clinical C5-neutralizing mAb treatment prevents the formation of cytolytic membrane attack complex (MAC) of complement, but does not block APC activation. Consequently, PNH patients are left with immune-mediated hemolytic anemia and their erythrocytes become opsonized with complement C3. Monoclonal antibodies (mAbs) against C3b/iC3b are useful for monitoring and studying C3b/iC3b deposit on PNH blood cells and mAbs with neutralizing activities are useful tools for studying C3-mediated CPC and APC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3b/iC3b/C3dg
Applications Flow Cytometry
Classification Monoclonal
Clone 1H8
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing PBS without preservatives.
Gene Accession No. P01024
Gene Symbols C3; CPAMD1
Host Species Mouse
Immunogen Sepharose 4B beads with surface C3b/C3bi deposits via APC in normal human serum corresponding to the iC3dg of human Complement C3b/iC3b/C3dg.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_000055
Target Species Human, Primate
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.