Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C3aR Antibody (17), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1496240.05MG
Description
Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Complement Component C3a R | |
Monoclonal | |
0.1 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, HNFAG09 | |
Mouse | |
Affinity Purified | |
RUO | |
719 | |
Human | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
17 | |
Unconjugated | |
0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide | |
C3AR1 | |
Human C3a Receptor | |
0.05 mg | |
Primary | |
The monoclonal antibody 17 reacts with the human receptor for C3a. The monoclonal antibody 17 is in the literature also referred to as clone hC3aRZ8. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction