Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS5, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89113409 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-409 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-409 Supplier Abnova Corporation Supplier No. MAB8267
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant COPS5.

Specifications

Antigen COPS5
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant COPS5.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Cops5
Gene Alias AI303502/CSN5/Jab1/Mov34/Sgn5
Gene Symbols Cops5
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-334 of mouse COPS5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26754
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.