Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CORIN, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106387 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-387 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-387 Supplier Abnova Corporation Supplier No. H00010699A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CORIN.

This gene encodes a member of the type II transmembrane serine protease class of the trypsin superfamily. Members of this family are composed of multiple structurally distinct domains. The encoded protein converts pro-atrial natriuretic peptide to biologically active atrial natriuretic peptide, a cardiac hormone that regulates blood volume and pressure. This protein may also function as a pro-brain-type natriuretic peptide convertase. [provided by RefSeq

Sequence: CKERDLWECPSNKQCLKHTVICDGFPDCPDYMDEKNCSFCQDDELECANHACVSRDLWCDGEADCSDSSDEWDCVTLSINVNSSSFLMVHRAATEHHVCA

Specifications

Antigen CORIN
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CORIN.
Formulation 50% glycerol
Gene CORIN
Gene Accession No. NM_006587
Gene Alias ATC2/CRN/Lrp4/MGC119742/TMPRSS10
Gene Symbols CORIN
Host Species Mouse
Immunogen CORIN (NP_006578, 616 a.a. to 715 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10699
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.