Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corneal epithelial keratins, Mouse, Clone: AE5, Abnova™

Catalog No. 89135193 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-193 Supplier Abnova Corporation Supplier No. MAB2449
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native rabbit corneal epithelial keratins.

Specifications

Antigen Corneal epithelial keratins
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone AE5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native rabbit corneal epithelial keratins.
Dilution Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified rabbit corneal epithelial keratins.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The 64K basic keratin K3.
Target Species Bovine, Human, Rabbit
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.