Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Coronin-1A, Clone: 2E4.1,

Catalog No. MABN319MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN319MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN319MI Supplier MilliporeSigma™ Supplier No. MABN319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Coronin-1A clone: 2E4.1 in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry

Coronin-1A (also known as clipin-A, coronin-1, coronin-like protein, Acoronin-like protein p57, tryptophan aspartate-containing coat protein) is a cytoplasmic protein encoded by the CORO1A gene in human (also known as CLABP, CLIPINA, HCORO1, IMD8, TACO). It is a member of the WD repeat protein family of proteins that are reported to be involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Coronin-1A is reported to be a component of the phagosome coat that is normally released prior to maturation into lysosomes. Mycobacteria are reported to evade host defense by recruiting and retaining coronin-1A on phagosomes, while in coronin-1A-lacking macrophages, mycobacteria are readily transported to lysosomes for degradation (PMID 10338208).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin-1A
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 2E4.1
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P31146
Gene Symbols CORO1A; CORO1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Coronin-1A.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001180262
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.