Learn More
COTL1, Mouse, Clone: 5C8, Abnova™
Mouse monoclonal antibody raised against partial recombinant COTL1.
Supplier: Abnova Corporation MAB10571
Description
This gene encodes one of the numerous actin-binding proteins which regulate the actin cytoskeleton. This protein binds F-actin, and also interacts with 5-lipoxygenase, which is the first committed enzyme in leukotriene biosynthesis. Although this gene has been reported to map to chromosome 17 in the Smith-Magenis syndrome region, the best alignments for this gene are to chromosome 16. The Smith-Magenis syndrome region is the site of two related pseudogenes. [provided by RefSeq
Specifications
COTL1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
COTL1 | |
COTL1 | |
Recombinant protein corresponding to human COTL1. | |
RUO | |
23406 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
5C8 | |
Mouse monoclonal antibody raised against partial recombinant COTL1. | |
In PBS (0.5% protein stabilizer, 0.05% sodium azide) | |
CLP/FLJ43657/MGC19733 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.