Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPNE7, Mouse, Clone: CPNE7-01, Abnova™

Catalog No. 89111495 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-495 Supplier Abnova Corporation Supplier No. MAB3603
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CPNE7.

This gene encodes a member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains and one von Willebrand factor A domain. The encoded protein may be involved in membrane trafficking. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CPNE7
Applications Western Blot
Classification Monoclonal
Clone CPNE7-01
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CPNE7.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CPNE7
Gene Alias MGC34192
Gene Symbols CPNE7
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to C-terminus human CPNE7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27132
Test Specificity This antibody recognizes C terminus of human CPNE7, a calcium-bindingcytoplasmic protein expressed mainly in brain.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.