Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CR2, Mouse, Clone: 4C5, Abnova™

Catalog No. 89112206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-206 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-206 Supplier Abnova Corporation Supplier No. MAB5288
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CR2.

Complement component receptor-2 (CR2) is the membrane protein on B lymphocytes to which the Epstein-Barr virus (EBV) binds during infection of these cells. See also CR1 (MIM 120620). Yefenof et al. (1976) [PubMed 181330] found complete overlapping of EBV receptors and C3 (MIM 120700) receptors on human B lymphocytes.[supplied by OMIM

Specifications

Antigen CR2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 4C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CR2.
Dilution Immunohistochemistry (Frozen sections) (1:20-1:50) Flow Cytometry (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CR2
Gene Alias C3DR/CD21/SLEB9
Gene Symbols CR2
Host Species Mouse
Immunogen Immunized subcutaneously in its footpads with fragments of human tonsil.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1380
Test Specificity This antibody reacts with the CD21 (140kD) molecule, expressed (moderate) on mature B-cells and (at high density) on follicular dendritic cells (FDC).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.