Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRABP1, Mouse, Clone: AT1A1, Abnova™

Catalog No. 89113184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-184 Supplier Abnova Corporation Supplier No. MAB7558
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant CRABP1.

This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue. [provided by RefSeq

Specifications

Antigen CRABP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1A1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant CRABP1.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CRABP1
Gene Alias CRABP/CRABP-I/CRABPI/RBP5
Gene Symbols CRABP1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-137 of human CRABP1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1381
Test Specificity This antibody reacts with human CRABP1.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.