Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRB1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-603 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-603 Supplier Abnova Corporation Supplier No. H00023418A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CRB1.

This gene encodes a protein which is similar to the Drosophila crumbs protein and localizes to the inner segment of mammalian photoreceptors. In Drosophila crumbs localizes to the stalk of the fly photoreceptor and may be a component of the molecular scaffold that controls proper development of polarity in the eye. Mutations in this gene are associated with a severe form of retinitis pigmentosa, RP12, and with Leber congenital amaurosis. Alternatively spliced transcript variants have been observed but their full-length nature has yet to be determined. [provided by RefSeq

Sequence: FCNKNNTRCLSNSCQNNSTCKDFSKDNDCSCSDTANNLDKDCDNMKDPCFSNPCQGSATCVNTPGERSFLCKCPPGYSGTICETTIGSCGKNSCQHGGICHQDPIYPVC

Specifications

Antigen CRB1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CRB1.
Formulation 50% glycerol
Gene CRB1
Gene Accession No. NM_012076
Gene Alias LCA8/RP12
Gene Symbols CRB1
Host Species Mouse
Immunogen CRB1 (NP_036208, 26 a.a. to 134 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23418
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.