Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB1, Mouse, Clone: B1C4, Abnova™

Catalog No. 89112861 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-861 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-861 Supplier Abnova Corporation Supplier No. MAB6423
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic phosphopeptide of CREB1.

This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen CREB1
Applications ELISA
Classification Monoclonal
Clone B1C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of CREB1.
Dilution ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.0
Gene CREB1
Gene Alias CREB/MGC9284
Gene Symbols CREB1
Host Species Mouse
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S133 of human CREB1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1385
Test Specificity Reactive to the immunogen, peptide-BSA conjugating and peptide in ELISA.
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.