Learn More
CREB5, Mouse, Clone: 1E2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CREB5.
Supplier: Abnova Corporation H00009586M01A
Description
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: MFCTSGGNSASVMSMRPVPGSLSSLLHLHNRQRQPMPASMPGTLPNPTMPGSSAVLMPMERQMSVNSSIMGMQGPNLSNPCASPQVQPMHSEAKMRLKASpecifications
| CREB5 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_001011666 | |
| CREB5 | |
| CREB5 (NP_001011666,1 a.a. ∽ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9586 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1E2 | |
| Mouse monoclonal antibody raised against a partial recombinant CREB5. | |
| CREB5 | |
| CRE-BPA | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.