Learn More
CREB5, Mouse, Clone: 7D4, Abnova™
Mouse monoclonal antibody raised against partial recombinant CREB5.
Supplier: Abnova Corporation MAB0819
Description
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
CREB5 | |
Monoclonal | |
Unconjugated | |
Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
CREB5 | |
CREB5 | |
Recombinant His fusion protein corresponding to amino acids 373-508 of human CREB5. | |
100 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
7D4 | |
Mouse monoclonal antibody raised against partial recombinant CREB5. | |
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide) | |
CRE-BPA | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
9586 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.