Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRP, Mouse, Clone: 2F12F6, Abnova™

Catalog No. 89110496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-496 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-496 Supplier Abnova Corporation Supplier No. MAB1564
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length native CRP.

Specifications

Antigen CRP
Applications ELISA
Classification Monoclonal
Clone 2F12F6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native CRP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Gene CRP
Gene Alias MGC149895/MGC88244/PTX1
Gene Symbols CRP
Host Species Mouse
Immunogen Native purified human CRP.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1401
Test Specificity This antibody is specific to CRP and has no cross reaction with other human serum proteins.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.