Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-CRTC1, Clone: 5F3.1,

Catalog No. MABE461MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE461MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE461MI Supplier MilliporeSigma™ Supplier No. MABE461
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CRTC1 clone: 5F3.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting

CREB-regulated transcription coactivator 1 (UniProt Q6UUV9; also known as Mucoepidermoid carcinoma translocated protein 1, TORC-1, Transducer of CREB protein 1, Transducer of regulated cAMP response element-binding protein 1) is encoded by the CRTC1 (also known as KIAA0616, MECT1, TORC1, WAMTP1) gene (Gene ID 23373) in human. CRTC1 belongs to a family of glycosylated Ser/Thr-rich phosphoproteins that bind the basic leucine zipper (bZIP) region of Ca2+/cAMP-responsive element-binding protein (CREB) and function as CREB coactivators. CRTCs participate in CREB-mediated transcriptions of both canonical cellular cAMP response elements (CREs) and the non-canonical viral CREs. CRTC1, specifically, is involved in CREB-mediated transcription of various neuronal immediate early genes (IEGs) as well as CREB-dependent hepatitis B virus (HBV) replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRTC1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5F3.1
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q6UUV9
Gene Symbols CRTC1; KIAA0616; MECT1; TORC1; WAMTP1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human CRTC1/TORC-1 sequence within the N-terminal half..
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001091952
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.