Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-CRYAA, Clone: 1H3.B8, Abnova™

Catalog No. 89110874 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-874 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-874 Supplier Abnova Corporation Supplier No. MAB2178
Add to Cart
Add to Cart

Specifications

Antigen CRYAA
Applications Western Blot
Classification Monoclonal
Clone 1H3.B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CRYAA.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene CRYAA
Gene Alias CRYA1/HSPB4
Gene Symbols CRYAA
Host Species Mouse
Immunogen Native purified CRYAA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1409
Test Specificity It does not cross-react with alpha B crystallin, beta-L crystallin, beta-H crystallin, gamma crystallin, Hsp25, Hsp27, or Hsp47.
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.