Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSAD, Mouse, Clone: 2C11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CSAD.
Supplier: Abnova Corporation H00051380M02A
Description
Sequence: MADSEALPSLAGDPVAVEALLRAVFGVVVDEAIQKGTSVSQKVCEWKEPEELKQLLDLELRSQGESQKQILERCRAVIRYSVKTGHPRFFNQLFSGLDSpecifications
CSAD | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_015989 | |
CSAD | |
CSAD (NP_057073,1 a.a. ∽ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
51380 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
2C11 | |
Mouse monoclonal antibody raised against a partial recombinant CSAD. | |
CSAD | |
CSD/FLJ44987/FLJ45500/MGC119354/MGC119355/MGC119357/PCAP | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction