Learn More
CSF1, Mouse, Clone: 116, Abnova™
Mouse monoclonal antibody raised against recombinant CSF1.
Supplier: Abnova Corporation MAB6295
Description
The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of macrophages. The active form of the protein is found extracellularly as a disulfide-linked homodimer, and is thought to be produced by proteolytic cleavage of membrane-bound precursors. The encoded protein may be involved in development of the placenta. Four transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq
Specifications
CSF1 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
CSF1 | |
CSF1 | |
Recombinant protein corresponding to human CSF1. | |
RUO | |
1435 | |
Human | |
Lyophilized |
ELISA | |
116 | |
Mouse monoclonal antibody raised against recombinant CSF1. | |
Lyophilized from 10mM PBS, pH 7.2 | |
MCSF/MGC31930 | |
Mouse | |
500 μg | |
Primary | |
This antibody is specifically reactive to CSF1. | |
Store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
|
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.