Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF2RA/B, Mouse, Clone: mxsghk-GMCSF, Abnova™

Catalog No. 89110342 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-342 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-342 Supplier Abnova Corporation Supplier No. MAB1305
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant CSF2RA/B.

Specifications

Antigen CSF2RA/B
Applications ELISA
Classification Monoclonal
Clone mxsghk-GMCSF
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant CSF2RA/B.
Dilution ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Alias CD116/CDw116/CSF2R/CSF2RAX/CSF2RAY/CSF2RX/CSF2RY/GM-CSF-R-alpha/GMCSFR/GMR/MGC3848/MGC4838
Gene Symbols CSF2RA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CSF2RA/B.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1438/1439
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.