Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF3, Mouse, Clone: GSB2, Abnova™

Catalog No. 89111181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-181 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-181 Supplier Abnova Corporation Supplier No. MAB2975

Mouse monoclonal antibody raised against recombinant CSF3.

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes. The active protein is found extracellularly. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CSF3
Applications ELISA
Classification Monoclonal
Clone GSB2
Conjugate Unconjugated
Description colony stimulating factor 3 (granulocyte)
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene CSF3
Gene Alias G-CSF, GCSF, MGC45931
Gene Symbols CSF3
Host Species Mouse
Immunogen Recombinant protein corresponding to human CSF3.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1440
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.