Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF3R, Mouse, Clone: S-425, Abnova™

Catalog No. 89112780 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-780 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-780 Supplier Abnova Corporation Supplier No. MAB6265
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant CSF3R.

The protein encoded by this gene is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble. Mutations in this gene are a cause of Kostmann syndrome, also known as severe congenital neutropenia. [provided by RefSeq

Specifications

Antigen CSF3R
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S-425
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant CSF3R.
Dilution Western Blot (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene CSF3R
Gene Alias CD114/GCSFR
Gene Symbols CSF3R
Host Species Mouse
Immunogen Recombinant MBP fusion protein corresponding to human CSF3R.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1441
Test Specificity This antibody is reactive with GSF3R and does not cross-react with MBP or BSA.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.