Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSK, Mouse, Clone: PAG-C1, Abnova™

Catalog No. 89111584 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-584 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-584 Supplier Abnova Corporation Supplier No. MAB3766
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of CSK.

Specifications

Antigen CSK
Applications Immunoprecipitation
Classification Monoclonal
Clone PAG-C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of CSK.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CSK
Gene Alias MGC117393
Gene Symbols CSK
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus last 15 amino acids of human CSK.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1445
Test Specificity This antibody recognizes an epitope located in the C-terminal domain of Csk-binding protein (Cbp).
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.