Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSNK2B, Mouse, Clone: 2, Abnova™

Catalog No. 89113855 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-855 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-855 Supplier Abnova Corporation Supplier No. MAB9897
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant CSNK2B.

This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus. [provided by RefSeq

Specifications

Antigen CSNK2B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant CSNK2B.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene CSNK2B
Gene Accession No. NM_001320.5
Gene Alias CK2B/CK2N/CSK2B/G5A/MGC138222/MGC138224
Gene Symbols CSNK2B
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CSNK2B.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1460
Test Specificity It can expression in HepG2 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.