Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSPG4, Mouse, Clone: LHM 2, Abnova™

Catalog No. 89113133 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-133 Supplier Abnova Corporation Supplier No. MAB7129
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CSPG4.

A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells. [provided by RefSeq

Specifications

Antigen CSPG4
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LHM 2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CSPG4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05 sodium azide)
Gene CSPG4
Gene Alias HMW-MAA/MCSP/MCSPG/MEL-CSPG/MSK16/NG2
Gene Symbols CSPG4
Host Species Mouse
Immunogen Native purified CSPG4 from A375P cell crude extract.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1464
Test Specificity MCSP (LHM 2). Predicted molecular weight 240 KDa.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.