Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CT45A3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CT45A3 protein.
Supplier: Abnova Corporation H00441519B01P
Description
Sequence: MTDKTEKVAVDPETVFKRPRECDSPSYQKRQRMALLARKQGAGDSLIAGSAMSKEKKLMTGHAIPPSQLDSQIDDFTGFSKDRMMQKPGSNAPVGGNVTSSFSGDDLECRETASSPKSQREINADIKRKLVKELRCVGQKYEKIFEMLEGVQGPTAVRKRFFESIIKEAARCMRRDFVKHLKKKLKRMISpecifications
CT45A3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human CT45A3 protein. | |
CT45A3 | |
CT45-3/CT45.3 | |
Mouse | |
Affinity chromatography | |
RUO | |
441519 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001017435.1 | |
CT45A3 | |
CT45A3 (NP_001017435.1,1 a.a. ∽ 189 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction