Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTRC Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89110896 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-896 Supplier Abnova Corporation Supplier No. MAB2210
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CTRC.

This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined. [provided by RefSeq

Specifications

Antigen CTRC
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CTRC.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene CTRC
Gene Alias CLCR/ELA4
Gene Symbols CTRC
Host Species Mouse
Immunogen Native purified CTRC from human pancreas.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11330
Test Specificity These antibodies do not cross-react with human pancreatic trypsin.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.