Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTSD, Mouse, Clone: 4G2, Abnova™

Catalog No. 89109033 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-033 Supplier Abnova Corporation Supplier No. MAB1083
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant CTSD.

This gene encodes a lysosomal aspartyl protease composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. This proteinase, which is a member of the peptidase C1 family, has a specificity similar to but narrower than that of pepsin A. Transcription of this gene is initiated from several sites, including one which is a start site for an estrogen-regulated transcript. Mutations in this gene are involved in the pathogenesis of several diseases, including breast cancer and possibly Alzheimer disease. [provided by RefSeq

Specifications

Antigen CTSD
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant CTSD.
Dilution Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene CTSD
Gene Accession No. NM_001909
Gene Alias CLN10/CPSD/MGC2311
Gene Symbols CTSD
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 21-412 of human CTSD.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1509
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.