Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTSL1, Mouse, Clone: CP-L14, Abnova™

Catalog No. 89110427 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-427 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-427 Supplier Abnova Corporation Supplier No. MAB1432
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CTSL1.

The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. At least two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen CTSL1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 33/2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CTSL1.
Dilution Western Blot (2 ug/mL) Immunohistochemistry (Frozen sections) (4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide)
Gene CTSL1
Gene Alias CATL/CTSL/FLJ31037/MEP
Gene Symbols CTSL1
Host Species Mouse
Immunogen Native purified human CTSL1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1514
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.