Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cycs, Mouse, Clone: 6H2.B4, Abnova™

Catalog No. 89112759 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-759 Supplier Abnova Corporation Supplier No. MAB6240
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cycs.

Specifications

Antigen Cycs
Applications Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 6H2.B4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cycs.
Dilution Immunoprecipitation (2 ug/106 cells) Immunohistochemistry (Frozen sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene Cycs
Gene Alias CYCSA/MGC93634
Gene Symbols Cycs
Host Species Mouse
Immunogen Rat Cycs.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25309
Test Specificity Clone 6H2.B4 (IMG-102A) recognizes the native form of Cytochrome c (Goshorn et al. 1991). Native cytochrome c has an approximate molecular weight of 15 KDa. Clone 6H2. B4 does not appear to recognize the denatured form of Cytochrome c. This antibody recognizes an epitope around residue 62 of rat Cytochrome c, based on copetitive ELISA results (Goshorn et al. 1991).
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.