Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyp1a1, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89111607 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-607 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-607 Supplier Abnova Corporation Supplier No. MAB3813
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Cyp1a1.

Specifications

Antigen Cyp1a1
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Cyp1a1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% gelatin, 0.05% sodium azide)
Gene Cyp1a1
Gene Alias AHH/AHRR/CP11/CYP1/Cyp45c/Cypc45c/P-450MC/P1-450/P450-C/P450DX
Gene Symbols Cyp1a1
Host Species Mouse
Immunogen Recombinant protein corresponding to rat Cyp1a1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24296
Test Specificity This mouse monoclonal antibody is specific to rat cytochrome P450 1A1 and is provided as IgG from caprylic acid and ammonium sulfate treatments. This monoclonal mouse IgG is useful for the identification of CYP450 1A1 by immunoassay or Western blots. Does not cross-react with 1A2.
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.