Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyp1a2, Mouse, Clone: 3B8C1, Abnova™

Catalog No. 89109308 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-308 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-308 Supplier Abnova Corporation Supplier No. MAB11116
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant protein of rat Cyp1a2.

Specifications

Antigen Cyp1a2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3B8C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant protein of rat Cyp1a2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.01% sodium azide)
Gene Cyp1a2
Gene Alias CYPD45/P-450d/RATCYPD45
Gene Symbols Cyp1a2
Host Species Mouse
Immunogen Recombinant protein corresponding to rat Cyp1a2.
Purification Method Protein A/G purification
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24297
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.