Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyp2b1/Cyp2b2, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89111608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-608 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-608 Supplier Abnova Corporation Supplier No. MAB3814
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cyp2b1/Cyp2b2.

Specifications

Antigen Cyp2b1/Cyp2b2
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cyp2b1/Cyp2b2.
Dilution Western Blot (1:100) The optimal working dilution should be determined by the end user.
Formulation In purified antibody
Gene Alias Cyp2b-1/Cypbe/MGC188065
Gene Symbols Cyp2b1
Host Species Mouse
Immunogen Rat Cyp2b1/Cyp2b2.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24300/361523
Test Specificity Identification of 2B1 and 2B2 by immunoassay or Western blots. This antibody also cross-reacts weakly with P450 2A1.
Target Species Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.